Ethylchlorformiat 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
118255000
118250025
ACRO118255000EA
1020
DKK
ACRO118255000
ACRO118250025
Ethylchlorformiat 99%
Ethylchlorformiat
Formel:
C₃H₅ClO₂ Kogepunkt: 94 °C (760 mmHg) Smeltepunkt: –81 °C Massefylde: 1,135 g/cm³ Flammepunkt: 10 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00000644 CAS-nummer: 541-41-3 EINECS: 208-778-5 UN nummer: 1182 ADR: 6.1, Merck Index: 12,03831 |
Specifikationer Testresultater
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3940 to 1.3960 (20°C, 589 nm) |
GC | ≤1.0 % (diethyl carbonate) |
Color scale | ≤50 APHA |
Free acid (HCl) | ≤0.1 % |
Methanol | ≤0.05 % |
Peroxides | ≤1 mg/kg H2O2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...