4,5-Dichlor-o-phenylendiamin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine

113640050 113640250
ACRO113640050EA 718 DKK
ACRO113640050 ACRO113640250
4,5-Dichlor-o-phenylendiamin 98%
4,5-Dichlor-o-phenylendiamin
Formel: C₆H₆Cl₂N₂
Molekylvægt: 177.03 g/mol
Kogepunkt: 305
Smeltepunkt: 162…163 °C
MDL Number: MFCD00007723
CAS-nummer: 5348-42-5
EINECS: 226-305-0

Bestil nu

Specifikationer Testresultater

Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR