Gas / Gas cabinet EI 30, in two sizes
Leverandør: JINVALL INREDNINGAR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EI30-GAS2 EI30-GAS1 EI30H12
765-0988EA 42600 DKK
765-0988 765-0987 765-0991
Gas / Gas cabinet EI 30, in two sizes
Møbler Opbevaringskabinetter
EI 30-Gas / Gasol cabinet is equipped with ventilation openings in the back. If the cabinet is placed against the outer wall, ventilation can take place through the outer wall without mechanical ventilation.
Ventilation openings: 2 pipe sockets (type DN32) Ø 42/32 mm placed on the back of the cabinet, one at the top and one at the bottom.