Torso with interchangeable sex
Leverandør: 3B Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1000196 1000197
761-0002EA 0 DKK
761-0002 763-0095
Torso with interchangeable sex
Modeller Humanbiologi modeller
Torso, 20 parts: Torso in life size. Detachable head (two parts), stomach (one part) and intestinal package (two parts). Interchangeable male and female genitals. A total of 20 parts. Parts: head with cerebral hemisphere, eye, thorax, two halves of the lungs, heart (in two parts), liver, colon with small intestine and appendix (in four parts), incision of a kidney, female and male genitals (in two and four parts respectively) . Torson is mounted on a base.
  • Also available without head
Torso, 24 parts: Interchangeable male and female genitals and detachable head. Parts: head (in 3 parts) with half of the brain, eye, thorax, two halves of the lungs, heart (in two parts), stomach (in 2 parts), digestive organs (in 4 parts) liver, colon with small intestine and appendix, incision of a kidney , female and male genitals (in two and four parts respectively). Life-size Torson is mounted on a base. 87 × 38 × 25 cm. 7.1 kg.