2,4-Pentandiol (±) + meso ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Purity: 99%
CAS No.: 625-69-4
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 101°(213°F)
Refractive Index: 1.4350
Odour: Odourless
Boiling Point: 200-201°
Beilstein Reference: 969186
MDL No.: MFCD00004549
Formel:
C₅H₁₂O₂ Kogepunkt: 200…201 °C Smeltepunkt: ∼53 °C Massefylde: 0,955 Flammepunkt: 101 °C (213 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004549 CAS-nummer: 625-69-4 EINECS: 210-907-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...