2,3,6-Trifluorphenol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC7817D-10G PC7817D-50G PC7817D-100G PC7817D-25G
APOSPC7817D-10GEA 676 DKK
APOSPC7817D-10G APOSPC7817D-50G APOSPC7817D-100G APOSPC7817D-25G
2,3,6-Trifluorphenol 98%
2,3,6-Trifluorphenol
Formel: C₆H₃F₃O
Smeltepunkt: 31…37 °C
Flammepunkt: 64 °C
MDL Number: MFCD00061214
CAS-nummer: 113798-74-6
UN nummer: 1325
ADR: 4.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR