4-fluorbenzamidin, HCl 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Fluorophenylformamidine hydrochloride
PC401011-1G
PC401011-5G
PC401011-10G
APOSPC401011-1GEA
404
DKK
APOSPC401011-1G
APOSPC401011-5G
APOSPC401011-10G
4-fluorbenzamidin, HCl 97%
4-fluorbenzamidin, HCl
Formel:
C₇H₈ClFN₂ Molekylvægt: 174.61 g/mol Smeltepunkt: 221 °C |
MDL Number:
MFCD04114421 CAS-nummer: 456-14-4 UN nummer: 2811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...