Acetohydroxamic acid tech

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Hydroxyacetamide

OR72063-25G OR72063-100G
APOSOR72063-25GEA 273 DKK
APOSOR72063-25G APOSOR72063-100G
Acetohydroxamic acid tech
Acetohydroxamic acid
Formel: C₂H₅NO₂
Kogepunkt: 231,4 °C
Smeltepunkt: 88 - 90 °C
MDL Number: MFCD00009994
CAS-nummer: 546-88-3
UN nummer: 3077
ADR: 9+EHS,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR