1,3,5-Triphenoxybenzene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR72112-50G OR72112-100G
APOSOR72112-50GEA 20740 DKK
APOSOR72112-50G APOSOR72112-100G
1,3,5-Triphenoxybenzene
1,3,5-Triphenoxybenzene
Formel: C₂₄H₁₈O₃
Smeltepunkt: 108…110 °C
MDL Number: MFCD00367787
CAS-nummer: 23879-81-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR