(2-Methoxyphenyl)eddikesyre 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methoxyphenylacetic acid

OR6815-100G OR6815-25G OR6815-500G
APOSOR6815-100GEA 327 DKK
APOSOR6815-100G APOSOR6815-25G APOSOR6815-500G
(2-Methoxyphenyl)eddikesyre 97%
(2-Methoxyphenyl)eddikesyre
Formel: (C₆H₄OCH₃)CH₂COOH
Smeltepunkt: 122…125 °C
MDL Number: MFCD00004321
CAS-nummer: 93-25-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR