(6-Bromo-2-methylquinolin-4-yl)methanol 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR930176-100MG OR930176-250MG
APOSOR930176-100MGEA 3260 DKK
APOSOR930176-100MG APOSOR930176-250MG
(6-Bromo-2-methylquinolin-4-yl)methanol 96%
(6-Bromo-2-methylquinolin-4-yl)methanol
Formel: C₁₁H₁₀BrNO
MDL Number: MFCD01416611
CAS-nummer: 885279-63-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR