2,4,5-Trifluorbenzonitril 94%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC99475-5G
PC99475-25G
PC99475-100G
APOSPC99475-5GEA
164
DKK
APOSPC99475-5G
APOSPC99475-25G
APOSPC99475-100G
2,4,5-Trifluorbenzonitril 94%
2,4,5-Trifluorbenzonitril
Formel:
C₇H₂F₃N Kogepunkt: 168…170 °C Massefylde: 1,373 Flammepunkt: 83 °C |
MDL Number:
MFCD00013289 CAS-nummer: 98349-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...