Transmitter Liquilines, 4-Wire CM14
Leverandør: Endress+Hauser
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CM14-1009/0 CM14-1020/0
ENDRCM14-1009/0EA 6040 DKK
ENDRCM14-1009/0 ENDRCM14-1020/0
Transmitter Liquilines, 4-Wire CM14
Elektroder Multiparameter Elektroder
Compact panel transmitter for a pH/ORP, conductivity or dissolved oxygen measuring point.
  • Simple commissioning and operation thanks to Memosens digital technology
  • Compact device in 48×96 mm panel-mounted housing - ideal for skid installations
The Liquiline CM14 is a 1-channel transmitter suitable for operating a standard measuring point in the food, power, water and wastewater industries. The compact device fits in the common cabinet cut-outs. The Memosens hot plug and play concept allows quick installation and configuration of digital conductivity, pH/ORP or dissolved oxygen sensors.
Leveringsinformation: Single channel transmitter Liquiline CM14.