Terbium 10.000 mg/l i fortyndet salpetersyre (from Tb4O7) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
456152TEA
4140
DKK
456152T
456154V
Terbium 10.000 mg/l i fortyndet salpetersyre (from Tb4O7) ARISTAR® enkeltelement standard til ICP
Standarder
Kationiske standarder
Terbium Standards
Tb₄O₇ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Certificeringer: Supplied with certificate of analysis.