7-Hydroxycoumarin-3-carboxylic acid ≥90%

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Umbelliferone-3-carboxylic Acid, 7-Hydroxy-2-oxo-2H-chromene-3-carboxylic acid

549 550
AATB549EA 1140 DKK
AATB549 AATB550
7-Hydroxycoumarin-3-carboxylic acid ≥90%
7-Hydroxycoumarin-3-carboxylic acid

7-Hydroxycoumarin-3-carboxylic acid is a blue fluorophores for labeling proteins and nucleic acids mostly through the in situ formation of its succinimidyl ester catalyzed by EDAC.

Formel: C₁₀H₆O₅
MDL Number: MFCD00017491
CAS-nummer: 779-27-1
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb3aa3996aa31d4dc7ce36803bfb5a398806300a&vid=2ba2fa32470a18fa8126cb6a4145b9f146e81560&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR