Fura 2-AM calcium indicator
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester
20494 20495
AATB20494EA 1720 DKK
AATB20494 AATB20495
Fura 2-AM calcium indicator
Fura 2-AM
Calcium measurement is critical for numerous biological investigations.
  • Cell permeable
  • Calcium-dependent fluorescence
Formel: C₄₄H₄₇N₃O₂₄
Molekylvægt: 1001.857 g/mol
MDL Number: MFCD00036976
CAS-nummer: 108964-32-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ce2dfdc6df90e35d4f28f262f061720b474a064&vid=f51999646dab9a7e5bec470ad3242aa804599680&prid=web&clid=SAPDAM