LDS 751 95
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: LDS-751, 6-(Dimethylamino)-2-(4-(4-(dimethylamino)phenyl)buta-1,3-dien-1-yl)-1-ethylquinolin-1-ium perchlorate
17561 17567
AATB17561EA 4240 DKK
AATB17561 AATB17567
LDS 751 95
LDS 751
LDS 751 is a fluorescent cell-permeant nucleic acid stain that can be well excited with 488 nm laser line although it has a peak excitation at ~543 nm on dsDNA.
Formel: C₂₅H₃₀N₃·ClO₄
Molekylvægt: 471.98 g/mol
MDL Number: MFCD00467069
CAS-nummer: 181885-68-7
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71cb14f42934b511f48919162927fdfa62e141cb&vid=47c994ea1e206994230d79183b7fa8de4d8e37bc&prid=web&clid=SAPDAM