iFluor® 790, Succinimidyl Ester
Leverandør: AAT BIOQUEST
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1368
71368
71524
71574
AATB1368EA
2300
DKK
AATB1368
AATB71368
AATB71524
AATB71574
iFluor® 790, Succinimidyl Ester
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- Useful For In-Vivo Imaging
Advarsel: For research use only.