6-FAM Alkyne ≥95%
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
134 956
AATB134EA 1720 DKK
AATB134 AATB956
6-FAM Alkyne ≥95%
6-FAM Alkyne
6-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Advarsel: For research use only.
CAS-nummer: 478801-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ad601d181a10648ba2cc21469b8521200599432&vid=aff8e6aee114db050f3c60fb78c7011c459c9204&prid=web&clid=SAPDAM