iFluor® 710, Succinimidyl Ester
Leverandør: AAT BIOQUEST
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1045
71045
71520
71570
AATB1045EA
2300
DKK
AATB1045
AATB71045
AATB71520
AATB71570
iFluor® 710, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- pH-Independent Signal
Advarsel: For research use only.