Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 700)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

16714 16839
AATB16714EA 1140 DKK
AATB16714 AATB16839
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 700)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Advarsel: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR