Anti-IgG Goat Polyclonal Antibody (iFluor® 568)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

16541 16777
AATB16541EA 1140 DKK
AATB16541 AATB16777
Anti-IgG Goat Polyclonal Antibody (iFluor® 568)
Antistoffer
iFluor® 568 is a bright red fluorescent dye.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Advarsel: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR