MagaDye™ 588 ddTTP
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17061 17065
AATB17061EA 10520 DKK
AATB17061 AATB17065
MagaDye™ 588 ddTTP
Nukleinsyrereagenser Sekventeringsreagenser
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Advarsel: For research use only.