CytoWatch™ QZ100 Monocyte Blocking Reagent
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37000 37001
AATB37000EA 940 DKK
AATB37000 AATB37001
CytoWatch™ QZ100 Monocyte Blocking Reagent
Assays Cellulær Assays
Some dye-labeled fluorescent antibody conjugates used in cell surface staining of live cells often exhibit non-specific binding in monocytes and macrophages.
  • Suitable for PBMC or whole blood
  • Minimizes background signal
  • No effect on lymphocytes, monocytes, or granulocytes
Advarsel: For research use only.