Cell Meter™ JC-10 Mitochondrion Membrane Potential Assay Kits
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22800 22801
AATB22800EA 3440 DKK
AATB22800 AATB22801
Cell Meter™ JC-10 Mitochondrion Membrane Potential Assay Kits
Assays Cellulær Assays
Although JC-1 is widely used in many labs, its poor water solubility causes extraordinary inconvenience.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Advarsel: For research use only.