Screen Quest™ Fluorimetric Glucose Uptake Assay Kits
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36500 36501
AATB36500EA 8140 DKK
AATB36500 AATB36501
Screen Quest™ Fluorimetric Glucose Uptake Assay Kits
Assays Cellulær Assays
Glucose transport systems are responsible for transporting glucose across cell membranes.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for microplate reader but suitable for all major fluorescence platforms
Advarsel: For research use only.