Screen Quest™ Fluorimetric Glucose Uptake Assay Kits
Leverandør: AAT BIOQUEST
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36500
36501
AATB36500EA
8140
DKK
AATB36500
AATB36501
Screen Quest™ Fluorimetric Glucose Uptake Assay Kits
Assays
Cellulær Assays
Glucose transport systems are responsible for transporting glucose across cell membranes.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimised for microplate reader but suitable for all major fluorescence platforms
Advarsel: For research use only.