PhosphoWorks™ Luminometric ATP Assay Kit, Bright Glow
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21610 21621
AATB21610EA 1720 DKK
AATB21610 AATB21621
PhosphoWorks™ Luminometric ATP Assay Kit, Bright Glow
Assays Cellulær Assays
Adenosine triphosphate (ATP) plays a fundamental role in cellular energenics, metabolic regulation and cellular signaling.
  • Extreme sensitivity
  • Detects as few as 10 cells/well
  • No mixing or separations required
Advarsel: For research use only.