Hose nipple deionsation cartridge
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D8901 D8902 D8905 D8904 D8822 D50220 D8903 D8908 D8911 D8921 D8922
171-0224EA 1230 DKK
171-0224 171-0223 171-0222 171-0225 BARND8822 BARND50220 BARND8903 BARND8908 BARND8911 BARND8921 BARND8922
Hose nipple deionsation cartridge
Filtre Filterpatroner
The cartridge is designed for non-pressure applications, it includes 1 cm barbed hose connections on each end. The 100% virgin polypropylene cartridge rinses to high quality quickly. A colour indicator shows when the resin cartridge is exhausted.
  • Same superior quality resins used in B-Pure cartridges
  • Contain dyes that change colour when resin is exhausted
  • Ideal for small volume applications; lower cost alternative