Human Protein A-Agarose Resin
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
55000 55005
AATB55000EA 1140 DKK
AATB55000 AATB55005
Human Protein A-Agarose Resin
Proteiner
Protein A agarose was prepared by covalently immobilizing protein A onto high-quality crosslinked beaded agarose.
  • High affinity
  • Targets igg antibodies
  • pH stable between 3 to 13
Advarsel: For research use only.