ReadiLink™ Biotin Nick Translation dsDNA Labeling Kit
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17470 17471
AATB17470EA 4060 DKK
AATB17470 AATB17471
ReadiLink™ Biotin Nick Translation dsDNA Labeling Kit
Proteinkoblings- og mærkningssystemer
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearized plasmid DNA.
  • Provides all necessary reagents
  • Adjustable ratios
  • Wide sample type compatibility
The resulted Biotin-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridization (FISH).
Emballering: Packaged in plastic vials.
Advarsel: For research use only.