Screen Quest™ Calbryte-590 Probenecid-Free and Wash-Free Calcium Assay Kit
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36200 36201 36202
AATB36200EA 3440 DKK
AATB36200 AATB36201 AATB36202
Screen Quest™ Calbryte-590 Probenecid-Free and Wash-Free Calcium Assay Kit
Assays Cellulær Assays
Calcium flux assays are the preferred methods for screening G protein coupled receptors (GPCRs) in drug discovery.
  • 100 to 250 times fluorescence increase once bound to calcium
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Brighter and more photostable signal
Advarsel: For research use only.