Screen Quest™ Membrane Potential Assay Kit, Orange Fluorescence
Leverandør: AAT BIOQUEST
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
35999
36000
36001
AATB35999EA
2300
DKK
AATB35999
AATB36000
AATB36001
Screen Quest™ Membrane Potential Assay Kit, Orange Fluorescence
Assays
Cellulær Assays
Membrane potential is the difference in voltage between the interior and exterior of a cell.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Advarsel: For research use only.