Screen Quest™ Membrane Potential Assay Kit, Orange Fluorescence
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
35999 36000 36001
AATB35999EA 2300 DKK
AATB35999 AATB36000 AATB36001
Screen Quest™ Membrane Potential Assay Kit, Orange Fluorescence
Assays Cellulær Assays
Membrane potential is the difference in voltage between the interior and exterior of a cell.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Advarsel: For research use only.