Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 750)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16506 16748
AATB16506EA 896 DKK
AATB16506 AATB16748
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 750)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins
Advarsel: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 750
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse