Anti-IgG Goat Polyclonal Antibody (iFluor® 633)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

16638 16808
AATB16638EA 896 DKK
AATB16638 AATB16808
Anti-IgG Goat Polyclonal Antibody (iFluor® 633)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Advarsel: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor™ 633
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR