Human Recombinant ASGR2
Leverandør: OriGeneSamlet vurdering
Gennemsntilig vurdering
This gene encodes a subunit of the asialoglycoprotein receptor. This receptor is a transmembrane protein that plays a critical role in serum glycoprotein homeostasis by mediating the endocytosis and lysosomal degradation of glycoproteins with exposed terminal galactose or N-acetylgalactosamine residues.
- Expression host: HEK293T
- Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol
Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.
The asialoglycoprotein receptor may facilitate hepatic infection by multiple viruses including hepatitis B, and is also a target for liver-specific drug delivery. The asialoglycoprotein receptor is a hetero-oligomeric protein composed of major and minor subunits, which are encoded by different genes. The protein encoded by this gene is the less abundant minor subunit. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Advarsel: For testing in cell culture applications, please filter before use. Note that you may experience some loss of protein during the filtration process.