6-NED NHS ester fluorescent probe

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 6-NED, SE

214 215
AATB214EA 2180 DKK
AATB214 AATB215
6-NED NHS ester fluorescent probe
6-NED NHS ester

5'-Labeled primer pairs include a fluorescently labeled oligo with a choice of dye on the 5' end, along with an unlabeled oligo, in each pair.


  • Amine-reactive probe

Molekylvægt: 644.47 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae4e85f1cf0542757b8f6195dce9b25f7cc60094&vid=ef78f104cfec615c685d76e34f52f01977ddccaa&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR