6-FAM Azide ≥95%
Leverandør: AAT BIOQUEST
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
133
955
AATB133EA
1720
DKK
AATB133
AATB955
6-FAM Azide ≥95%
6-FAM Azide
6-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group.
- Multiple Bioconjugation Applications
- Excellent Quality
Advarsel: For research use only.
CAS-nummer:
1386385-76-7 |