5-FAM azide ≥95%
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
131 950
AATB131EA 1720 DKK
AATB131 AATB950
5-FAM azide ≥95%
5-FAM azide
5-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Advarsel: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7aac07f5ffe8cc3a0f4e03febde23801415192a&vid=6457ed13074836d995c602234654e221eb07b914&prid=web&clid=SAPDAM