Pipette washer/rinser, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5245-0040 5245-0050
NALG5245-0040EA 4260 DKK
NALG5245-0040 612-1829
Pipette washer/rinser, Nalgene®
Pipetter Pipettevasker
For washing and rinsing of pipettes. Insert a filled pipette basket. Attach it to your sink water or DI tap and drain for repeated, automatic fill and empty cycling. The cycles continue until water source is turned off.
  • One piece, leakproof, weld-free and made of robust HDPE
  • Large tubing diameter permits faster cycles and allows water to drain completely
  • Water inlet designed to eliminate back siphoning of contaminated water into water line, even if pressure drops
  • Slower cycling is achieved without attachments by turning down water source flow rate
  • Work well on low water pressure
Bestillingsoplysninger: Baskets are sold separately.