Galileo thermometers
Leverandør: MOLLER THERM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
106322 106323 106301 106333 106334 106321 106331 106304 106320 106350
765-1084EA 0 DKK
765-1084 765-1085 765-1080 765-1087 765-1088 765-1083 765-1086 765-1081 765-1082 765-1089
Galileo thermometers
Termometre Galileo Thermometers
Room thermometer with five (340 mm) or seven (420 mm) balls.
  • Fluid's density changes with temperature
  • Engraved weights hang in the partially fluid-filled balls
  • Reading at the bottom of the balls that float up into the cylinder
Colour in the ordering table indicates the colour of the balls.