Funnel holders
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
148500 148502
241-0343EA 815 DKK
241-0343 241-0344
Funnel holders
Retortstativer
PP, white.
To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8 to 14 mm with a locking nut.