Spectrum lamps with Pico 9 base
Leverandør: OSRAM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4050300210353 4050300212210 4050300212234 4050300210377
765-1126EA 8660 DKK
765-1126 765-1128 765-1129 765-1127
Spectrum lamps with Pico 9 base
Lyskilder Spektrallamper
For the production of monochromatic light together with suitable filters or for line spectra experimentation. The spectral lamps may only be connected to 220V AC via the choke.
Operating current: 1 A
Burn mode: Vertically standing
Socket: Pico 9