Distillation cooler, Liebig
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.700.400 1.700.250
201-0200EA 665 DKK
201-0200 201-0199
Distillation cooler, Liebig
Svaler
Without grinding.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Certificeringer: DIN 12576