AERO CHROME* Crew Performance Surgical Gowns
Leverandør: Halyard
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48392 48393 48394 48399 48400 48401
HALY48392EA 1800 DKK
HALY48392 HALY48393 HALY48394 HALY48399 HALY48400 HALY48401
AERO CHROME* Crew Performance Surgical Gowns
Engangsbeklædning
AERO CHROME* CREW Breathable surgical gowns combine unprecedented softness and breathability with maximum protection against fluids and pathogens.
  • Meets AAMI Level 4 standard, providing the highest level of both fluid and microbial protection in the critical zones
  • Proprietary fabric technology allows moisture vapor to escape while maintaining a high performance barrier
  • Designed to keep your OR staff both cool and protected during long, fluid-intensive procedures
  • Highest rating for resistance to ignition from surgical lasers
  • Excellent abrasion and lint resistance
Certificeringer: Meets ASTM F1671 in the critical zones (ties, fabric, sleeve seams), per AAMI PB70:2012 Level 4, Meets AAMI Level 4, AAMI PB70:2012, EN13795 High Performance, ASTM F1671, MOCON and cup crush testing, ISO 22610 (wet),ISO 22612 (dry), ASTM F1671 and ASTM F1670, Hydrohead-20811, Per ISO 11810, Per EN Resistance to Linting and Particulates test data. Abrasion resistant per Martindale test method. Meets ASTM D4966.