POL Green Industry Powerful Ecological Degreasing Cleaners
Leverandør: Basan
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2210317 2210323 2210369 2210395
BSNC2210317EA 1380 DKK
BSNC2210317 BSNC2210323 BSNC2210369 BSNC2210395
POL Green Industry Powerful Ecological Degreasing Cleaners
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Powerful concentrated eco-labelled degreaser for intensive cleaning of mineral and vegetable oils and fats, as well as rubber residue (sports facilities) and rubber on all surfaces: tiles, rough concrete, quartz concrete, epoxy or polyurethane paint, epoxy mortar, etc. Food contact approved.
  • pH: >12,5
  • pH in solution of 1%: 10,0 ±0,5
  • Dosage: 1 to 20%
  • Colour: Colourless to yellow
Can be used for steeping and impregnation of the surfaces with the solution with the dosage recommended below with mechanical cleaning action (scrubber dryer - low- and high-pressure systems - spray). Can also be applied in automatic scrubber without soaking - equip the machine according to the type of floor (discs or brushes). Sports hall floors: apply a 2% dilution with a scrubber-dryer and red pads to remove gum residues and resins.
In application with high-pressure systems, one is always advised to soak or impregnate the surfaces beforehand by means of a spray and to make use of the high pressure as a mechanical water action only.
Storage: Keep the product in its original closed packaging. Protect against freezing temperatures.
Dosage:
Daily and conventional application: 1%
Gum residue removal: 2%
Spray application: 5%
Occasional reconditioning: 5%
Eliminating oil stains through impregnation: 20%
Certificeringer: Certified with a European ecolabel.