Erlenmeyer-kolber med bafler ikke og med membranskruelåg, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
212833655 212834454 212835459 212834405
214-0234EA 245 DKK
214-0234 214-0235 214-0340 392-0393
Erlenmeyer-kolber med bafler ikke og med membranskruelåg, DURAN®
Kolber Erlenmeyer-kolber
DURAN®, borosilicate glass 3.3, clear, with or without PP screw cap plus PTFE membrane.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Four baffles create turbulent flow, increasing the surface of the liquid and allowing greater gas exchange when compared to flat bottomed Erlenmeyer flasks. Complete with or without 0,2 μm PTFE membrane screw cap (GL 45) both cap and flask are autoclavable. With graduations and white labelling area.
Med trykt sporingskode (batchidentifikation).