Pure-Fit® SIB® Smooth Inner Bore Connector Systems
Leverandør: Saint Gobain Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PFTC750PP PFR250X500PPAF PF135250PPAF PF135375PPAF PF135500PPAF PFC500PPAF PFLS250PPAF PFLS375PPAF PFLS500PPAF PFMS250PPAF PFMS375PPAF PFMS500PPAF PFT250PPAF PFT375PPAF PFT500PPAF PFX250PPAF PFX375PPAF PFY250PPAF PFY375PPAF PFY500PPAF
VERNPFTC750PPEA 0 DKK
VERNPFTC750PP VERNPFR250X500PPAF VERNPF135250PPAF VERNPF135375PPAF VERNPF135500PPAF VERNPFC500PPAF VERNPFLS250PPAF VERNPFLS375PPAF VERNPFLS500PPAF VERNPFMS250PPAF VERNPFMS375PPAF VERNPFMS500PPAF VERNPFT250PPAF VERNPFT375PPAF VERNPFT500PPAF VERNPFX250PPAF VERNPFX375PPAF VERNPFY250PPAF VERNPFY375PPAF VERNPFY500PPAF
Pure-Fit® SIB® Smooth Inner Bore Connector Systems
Fittings og stik
Pure-Fit SIB eliminates entrapment and leak points that can occur with traditional assembly systems. The unique internal design eliminates voids or dead space that can lead to turbulence or stagnation of fluid flow creating the potential for bacterial growth.
  • Total smooth inner bore (SIB) transition design
  • Eliminates entrapment and leak points that occur with traditional fitting systems
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilises the BarbLock® retention and seal system
  • Compatible with Saint-Gobain Sani-Tech®, C-Flex® and PharMed® flexible tubing
  • PP is gamma stable
  • All materials completely animal-free and BPA-free
  • Manufactured and packaged in a certified clean room
  • Validation guide available
Pure-Fit® SIB® fittings are fully compatible with Pure-Fit and BarbLock® retainers that provide complete 360° compression for leak-free connections. It is ideal for applications such as single-use assemblies, biopharmaceutical manufacturing, aseptic and sterile fill lines, filtration, general laboratory, final fill, product sampling and fluid transfer. Also available in autoclavable and gamma stable PVDF, please see separate product page.
Hardness: 85 (Shore D)
Certificeringer: USP Class VI and ISO criteria and have been physically tested to meet the most demanding applications and stringent test protocols.