Safety spectacles, uvex super fit
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9178.065 9178.315 9178.415 9178.286
111-0580EA 249 DKK
111-0580 111-0581 UVEX9178.415 UVEX9178.286
Safety spectacles, uvex super fit
Briller
These spectacles with ultra-thin wrap-around lens provide excellent protection against mechanical hazards.
  • Extra slim wrap-around lenses with a 6,5 base curve
  • Maximum optical clarity thanks to extremely thin wrap-around lens
  • With hinged side arms
  • Ultra light
Models with ETC coating are suitable for very humid atmospheres and environments with extreme temperature conditions.
Weight: 21 g
Certificeringer: EN 166, EN 170