Sterilisation Wraps for Steam, ETO or Gamma, Blue, Heavy Duty
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BHDSHEET1212 BHDSH01.5X01.5 BHDSHEET0202 BHDSHEET0303 BHDSHEET0404 BHDSHEET0606 BHDSHEET0909 BHDSHEET1515 BHDSHEET1818 BHDSHEET2020 BHDSHEET2424 BHDSHEET3030 BHDSHEET3636 BHDSHEET4040 BHDSHEET4545 BHDSHEET5454 BHDSHEET5472 BHDSHEET0808 4856BHD
129-2808EA 3450 DKK
129-2808 129-2801 129-2802 129-2803 129-2804 129-2805 129-2807 129-2809 129-2810 129-2811 129-2812 129-2815 129-2816 129-2817 129-2818 129-2819 129-2820 129-2806 KACC4856BHD
Sterilisation Wraps for Steam, ETO or Gamma, Blue, Heavy Duty
Steriliseringsindpakning
When secured with autoclave tape or elastic bands, these blue BHD wrappers provide an effective microbial barrier from external sources of contamination.
  • Designed for steam sterilisation; but also compatible with ETO or gamma irradiation
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibres bonded with a resin binder
  • Sufficient microbial barrier
  • Superior bacterial barrier properties compared to linen
  • Sterilisation tapes adhere firmly and remove easily
  • Ideal choice for light trays, small basins and towels
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO certified cleanrooms
These versatile wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
Certificeringer: Certificates of Conformance.