Pre-Sterilised Media Plate Transport and Incubation Bags
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SBBMGTVKSS0608 SBBMGTVKSS0810 SBBMGTVKSS1012 SBBMGTVKSS1617
129-2904EA 10600 DKK
129-2904 129-2905 129-2908 129-2909
Pre-Sterilised Media Plate Transport and Incubation Bags
Poser Steriliseringsposer
Eliminate the risk and expense of investigating the root causes of false positives with Keystone Cleanroom Products Sterile media plate transport and incubation bag system.
  • Ready to use
  • Validated sterile
  • Tyvek/Mylar packaging utilises hang hooks on each corner to facilitate use in isolators
  • Tyvek packaging is compatible with VHP decontamination cycles
  • Manufactured and packaged in an ISO certified cleanroom
  • Self-sealing Tyvek Peel pouches hold the media plates that are collected inside the isolator
  • These Tyvek pouches are transferred to the QC Lab
  • The media plates do not need to be removed from the pouches for incubation
The system provides complete protection from the time the plates are being removed from inside the isolator all the way through incubation. Custom packaging and configurations are also available please ask your Avantor representative for more information.
Leveringsinformation: Certificates of Sterility and Conformance with every shipment.
Emballering: 10 per pack, double pouched in Tyvek peel pouches.